Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90.1 Antibody (HIS51) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200312500UG
Description
CD90.1 Monoclonal specifically detects CD90.1 in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
CD90.1 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
thy-1 membrane glycoprotein, CD90, Thy-1 cell surface antigen | |
Mouse | |
Affinity Purified | |
RUO | |
7070 | |
Store at 4C. Do not freeze. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HIS51 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
THY1 | |
The immunogen for this antibody is CD90.1 (Thy-1.1, Thy1.1). | |
500 μg | |
Primary | |
Mouse, Rat | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction