Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.1 (Thy-1.1) Monoclonal Antibody (OX-7), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544198
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4198 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4198 Supplier Leinco Technologies Inc Supplier No. C851100MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Clone OX-7 activity is directed against both rat and murine CD90/Thy-1. Endotoxin Level: <1.0 EU/μg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD90.1 (Thy-1.1) Monoclonal antibody specifically detects CD90.1 (Thy-1.1) in Mouse, Rat samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Inhibition Assays

Specifications

Antigen CD90.1 (Thy-1.1)
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Inhibition Assays
Classification Monoclonal
Clone OX-7
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Thy1
Gene Accession No. P01830, P01831
Gene Alias CD7; CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen
Host Species Mouse
Immunogen Rat Thy1 antigen
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21838, 24832
Target Species Mouse, Rat
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.