Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90.2 Antibody (30-H12), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP128040
Description
CD90.2 Monoclonal specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay.Specifications
CD90.2 | |
Monoclonal | |
0.1 mg/mL | |
Flow Cytometry <= 0.2 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay | |
THY1 | |
Mouse thymus or spleen | |
0.1 mg | |
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
7070 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay | |
30-H12 | |
APC | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse CD90.2/Thy-1.2 alloantigen, Mr 18 kDa | |
Store at 4C. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction