Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.2 Rat anti-Mouse, APC, Clone: 53-2.1, BD

Catalog No. BDB561974 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561974 25 μg
BDB553007 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561974 Supplier BD Biosciences Supplier No. 561974
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 53-2.1 monoclonal antibody specifically binds to the CD90.2 (Thy-1.2) alloantigen on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), epithelial cells, fibroblasts, neurons, hematopoietic stem cells, but not B lymphocytes, of most mouse strains. The 53-2.1 antibody has been reported not to crossreact with Thy-1.1 (e.g., AKR/J, PL), or with rat Thy-1. CD90 is a glycosyolphosphatidylinositol-anchored membrane glycoprotein of the Ig superfamily that is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of thymocytes to thymic stroma. The 53-2.1 antibody has been reported to block the binding of anti-mouse CD90.2 clone 30-H12 (Cat. No. 553009) to immobilized thymocyte membranes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90.2
Applications Flow Cytometry
Classification Monoclonal
Clone 53-2.1
Concentration 0.2mg/mL
Conjugate APC
Description Thy-1.2; T25; Thymus cell antigen 1, theta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Thymus / Spleen
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.