Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | CD90.2 (Thy-1.2) |
---|---|
Clone | 53-2.1 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M028T03Y04A
![]() |
Invitrogen™
M028T03Y04A |
100 μg |
Each for $375.00
|
|
|||||
M028T02Y04A
![]() |
Invitrogen™
M028T02Y04A |
25 μg |
Each for $194.00
|
|
|||||
Description
CD90.2 (Thy-1.2) Monoclonal antibody specifically detects CD90.2 (Thy-1.2) in Mouse samples. It is validated for Flow CytometrySpecifications
CD90.2 (Thy-1.2) | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
P01831 | |
21838 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
53-2.1 | |
Flow Cytometry | |
NovaFluor Yellow 660 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
IgG2a κ | |
Affinity chromatography | |
Thy1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title