Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD90 Monoclonal Antibody (5a-8), Biotin
Mouse Monoclonal Antibody
$309.00 - $602.50
Specifications
Antigen | CD90 |
---|---|
Clone | 5a-8 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517745
|
Invitrogen™
MA517745 |
100 μg |
Each for $309.00
|
|
|||||
PIMA517746
|
Invitrogen™
MA517746 |
300 μg |
Each for $602.50
|
|
|||||
Description
CD90 Monoclonal Antibody for IHC (F), Flow
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.Specifications
CD90 | |
0.1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen) | |
Biotin | |
Mouse | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P01831 | |
21838 | |
CBA/J. | |
Antibody |
5a-8 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Mouse | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Thy1 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title