Learn More
Invitrogen™ CD90 Monoclonal Antibody (OX-7), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA180648
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
Specifications
CD90 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
P01830, P01831 | |
Thy1 | |
Rat Thy1 antigen. | |
100 μg | |
Primary | |
Mouse, Rat, Rabbit, Guinea Pig | |
Antibody | |
IgG1 |
Flow Cytometry | |
OX-7 | |
FITC | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Mouse | |
Protein A | |
RUO | |
100339279, 100379563, 21838, 24832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.