Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90 Mouse anti-Human, BUV395, Clone: 5-E10, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566219
Description
CD90 Monoclonal antibody specifically detects CD90 in Baboon, Cynomolgus, Dog, Human, Pig, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| CD90 | |
| Monoclonal | |
| Brilliant Ultraviolet 395 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Cell Biology | |
| Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey | |
| Purified |
| Flow Cytometry | |
| 5-E10 | |
| Vol. Per test: 5μL | |
| THY1; Thy-1 antigen; Thy-1 membrane glycoprotein | |
| Human HEL Cell Line | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
| IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction