Learn More
CD90/Mouse CD90.1 Mouse anti-Mouse, Rat, BUV496, Clone: OX-7 (also known as OX7), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD90/Mouse CD90.1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OX-7 (also known as OX7) |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Rat Thy-1; Mouse Thy-1.1 |
| Host Species | Mouse |
| Immunogen | Rat Thymocyte Thy-1 Antigen |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.