Learn More
Invitrogen™ CD90 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595007
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Liver Tissue, Rat Thymus Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
Specifications
CD90 | |
Polyclonal | |
Unconjugated | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21838, 24832, 7070 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P01830, P01831, P04216 | |
Thy1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human CD90 (20-39aa QKVTSLTACLVDQSLRLDCR). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.