Learn More
CD90 (Thy-1) Monoclonal Antibody (eBio5E10 (5E10)), Brilliant Violet™ 480, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$160.65 - $370.65
Specifications
Antigen | CD90 (Thy-1) |
---|---|
Clone | eBio5E10 (5E10) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
41-409-0941
![]() |
Invitrogen™
414090941 |
25 Tests |
Each
for $160.65
Save 3% Reg : |
|
|||||
41-409-0942
![]() |
Invitrogen™
414090942 |
100 Tests |
Each
for $370.65
Save 4% Reg : |
|
|||||
Description
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.Specifications
CD90 (Thy-1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Thy1 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Thy1 |
eBio5E10 (5E10) | |
Flow Cytometry | |
Brilliant Violet 480 | |
Mouse | |
Human | |
P04216 | |
7070 | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.