Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90 (Thy-1) Mouse anti-Human, RB545, Clone: 5E10, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569759
Description
CD90 (Thy-1) Monoclonal antibody specifically detects CD90 (Thy-1) in Human samples. It is validated for Flow cytometry
Specifications
CD90 (Thy-1) | |
Monoclonal | |
RB545 | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein | |
Human HEL Cell Line | |
25 Tests | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
5E10 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
7070 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction