Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90/Thy1 Antibody (OX-7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
Supplier: Novus Biologicals NB10065543SS
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
CD90/Thy1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 35383427) | |
P01830 | |
THY1 | |
Rat Thy 1 antigen | |
Primary | |
CD90 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
OX-7 | |
Unconjugated | |
PBS with 0.02% sodium azide | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Mouse | |
0.05 mg | |
7070 | |
Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction