Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD90/Thy1 Antibody (Thy-1A1), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2067F
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD90/Thy1 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
Thy-1A1 | |
Flow Cytometry, CyTOF-ready | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Human CD90/Thy1 | |
0.1 mL | |
Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
7070 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction