Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD90/Thy1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB20671T100UG
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.Specifications
CD90/Thy1 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
Chinese Hamster Ovary cell line, CHO-derived human CD90/THY1, Met1-Ser141, Accession No. P04216 | |
100 μg | |
7070.0 | |
Human | |
Supernatant |
Flow Cytometry | |
783922 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P04216 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human CD90/THY1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction