Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD91 |
---|---|
Clone | A2MR-a2 |
Concentration | 0.4 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H073T03B06A
![]() |
Invitrogen™
H073T03B06A |
100 Tests |
Each for $430.00
|
|
|||||
H073T02B06A
![]() |
Invitrogen™
H073T02B06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD91 Monoclonal antibody specifically detects CD91 in Human samples. It is validated for Flow CytometrySpecifications
CD91 | |
0.4 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
Q07954 | |
4035 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
A2MR-a2 | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
A2MR; AI316852; alpha 2-macroglobulin receptor; alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; LDL receptor related protein 1; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; Lrp; Lrp1; LRP-1; LRP1A; LRP-515; LRP-85; LRPICD; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; TGFBR5; type V tgf-beta receptor | |
IgG1 κ | |
Affinity chromatography | |
LRP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title