Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD91 Monoclonal Antibody (A2MR-a2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$461.00
Specifications
Antigen | CD91 |
---|---|
Clone | A2MR-a2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
63-091-942
![]() |
Invitrogen™
63091942 |
100 Tests |
Each of 1 for $461.00
|
|
|||||
Description
Mouse Monoclonal Antibody
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.Specifications
CD91 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
4035 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
A2MR-a2 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
Q07954 | |
LRP1 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
LRP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title