Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD91 Mouse anti-Human, Brilliant Violet 421, Clone: A2MR-α2, BD Optibuild™

Catalog No. BDB743856 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743856 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743856 Supplier BD Biosciences Supplier No. 743856
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A2MR-α2 monoclonal antibody specifically reacts with a 600 kDa, type I membrane single protein, also known as theα2 Macroglobulin (α2M) receptor/low density lipoprotein receptor-related protein 1 (LRP-1). Reported to be an endocytic receptor involved with intracellular signalling, lipid homeostasis, clearance of apoptotic cells, andα2 Macroglobulin mediated clearance of secreted amyloid precursor protein found in Alzheimer patients. The single chain receptor undergoes cleavage, shortly after synthesis, into the 85 kDa transmembraneβchain that non-covalently binds to the extracellular 500-515 kDa a chain. It has a broad cellular distribution, but in the hematopoietic system it is expressed on monocyte lineage cells.α2M/LRP-1 mediates endocytosis of a variety of ligands includingα2M-proteinase complexes, plasminogen activators in complex with plasminogen activator inhibitor, or Pseudomonas /Exotoxin A. Ligand binding toα2M/LRP-1 is followed by rapid transport of the ligand to lysosomes for degradation.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD91
Classification Monoclonal
Clone A2MR-α2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias A2MR, APOER, APR, IGFBP3R, TGFBR5, LRPA, LRP1
Host Species Mouse
Immunogen Purified Human α2 Macroglobulin
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.