Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD91 Mouse anti-Human, BUV496, Clone: A2MR-α2, BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 750156
Description
CD91 Monoclonal antibody specifically detects CD91 in Human samples. It is validated for Flow Cytometry
Specifications
CD91 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
A2MR; APOER; APR; IGFBP3R; TGFBR5; LRPA; LRP1 | |
Purified Human α2 Macroglobulin | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
A2MR-α2 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction