Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD91 Monoclonal Antibody (A2MR-a2), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 46091942

Encompass_Preferred

 View more versions of this product

Catalog No. 50-113-0768


Only null left
Add to Cart

Description

Description

The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.

CD91 is a 600 kDa type I transmembrane protein composed of an alpha and beta chain. The alpha chain interacts with a variety of structurally and functionally diverse ligands, while the beta chain contains two tyrosine phosphorylation motifs that interact with adaptor and signaling proteins. This receptor is expressed by cells in the monocytic lineage, dendritic cells, hepatocytes, fibroblasts, adipocytes, neurons, and syncytiotrophoblasts. CD91 plays a crucial role in several cellular processes, including ligand endocytosis and trafficking to lysosomes, intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. It is involved in cross-presenting antigens via MHC class I molecules on antigen-presenting cells after endocytosis of heat shock protein-peptide complexes. Additionally, CD91 is essential for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of CD91 decreases with age and is found to be lower in brain tissue from Alzheimer patients compared to controls. The receptor also plays a role in cell migration and activating signaling cascades, highlighting its importance in both immune function and disease processes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD91
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q07954
LRP1
Affinity chromatography
RUO
4035
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
A2MR-a2
PerCP-eFluor 710
LRP1
A2MR; AI316852; alpha 2-macroglobulin receptor; alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; b2b1554Clo; CD91; FLJ16451; IGFBP3R; LDL receptor related protein 1; lipoprotein receptor-related protein; low density lipoprotein receptor-related protein 1; low density lipoprotein-related protein 1 (alpha-2-macroglobulin receptor); Low-density lipoprotein receptor-related protein 1 515 kDa subunit; Low-density lipoprotein receptor-related protein 1 85 kDa subunit; Low-density lipoprotein receptor-related protein 1 intracellular domain; Lrp; Lrp1; LRP-1; LRP1A; LRP-515; LRP-85; LRPICD; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; TGFBR5; type V tgf-beta receptor
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.