Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD91 Mouse, anti-Human, R718, Clone: A2MR-A2, BD OptiBuild™

Catalog No. BDB752150 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752150 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752150 Supplier BD Biosciences Supplier No. 752150
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A2MR-α2 monoclonal antibody specifically reacts with a 600 kDa, type I membrane single protein, also known as the α2 Macroglobulin (α2M) receptor/low density lipoprotein receptor-related protein 1 (LRP-1). Reported to be an endocytic receptor involved with intracellular signalling, lipid homeostasis, clearance of apoptotic cells, and α2 Macroglobulin mediated clearance of secreted amyloid precursor protein found in Alzheimer patients. The single chain receptor undergoes cleavage, shortly after synthesis, into the 85 kDa transmembrane β chain that non-covalently binds to the extracellular 500-515 kDa a chain. It has a broad cellular distribution, but in the hematopoietic system it is expressed on monocyte lineage cells. α2M/LRP-1 mediates endocytosis of a variety of ligands including α2M-proteinase complexes, plasminogen activators in complex with plasminogen activator inhibitor, or Pseudomonas Exotoxin A. Ligand binding to α2M/LRP-1 is followed by rapid transport of the ligand to lysosomes for degradation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD91
Applications Flow Cytometry
Classification Monoclonal
Clone A2MR-A2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.