Learn More
Invitrogen™ CD93 (AA4.1) Monoclonal Antibody (R3), PE, eBioscience™
Mouse Monoclonal Antibody
$353.00
Specifications
Antigen | CD93 (AA4.1) |
---|---|
Clone | R3 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-245-281
![]() |
Invitrogen™
12093842 |
100 Tests |
Each of 1 for $353.00
|
|
|||||
Description
Description: The monoclonal antibody R3 recognizes human CD93, also known as C1qRp. The glycoprotein CD93 binds to C1q, the subunit of the complement protein, mannose binding lectin and pulmonary surfactant protein A. CD93 is predicted to play a role in the clearance of apoptotic cells. Expression of CD93 is confined to myeloid cells with higher expression on monocytes than neutrophils, eosinophils, platelets and endothelial cells. Expression on DC's is downregulated upon maturation. Additionally, CD93 has been shown to define an early bone marrow stem cell population of hematopoietic and hepatic precursors. The R3 antibody has been shown to have activating function by increasing phagocytosis when presented in an immobilized state and inhibitory functions when presented in a soluble state. CD93 can be shed from the cell surface. This phenomenon can be measures using R3 antibody as detection with R139 as capture to detect soluble CD93 by ELISA. The epitope for R3 lies in the CRD domain.\\The R3 antibody has been shown to have activating function by increasing phagocytosis when presented in an immobilized state and inhibitory functions when presented in a soluble state. CD93 can be shed from the cell surface. This phenomenon can be measures using R3 antibody as detection with R139 as capture to detect soluble CD93 by ELISA. The epitope for R3 lies in the CRD domain. Applications Reported: This R3 antibody has been reported for use in flow cytometric analysis. Applications Te...
The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.Specifications
CD93 (AA4.1) | |
5 μL/Test | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q9NPY3 | |
22918 | |
Primary | |
Affinity chromatography |
R3 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgM | |
Human | |
Cd93 | |
6030404G09Rik; AA145088; Aa4; AA4.1; AW555904; C1q receptor; C1q receptor 1; C1q/MBL/SPA receptor; C1qR; C1qR(P); C1qr1; C1qRP; Cd93; CD93 antigen; CD93 molecule; CDw93; Cell surface antigen AA4; complement component 1 q subcomponent receptor 1; complement component 1, q subcomponent, receptor 1; complement component C1q receptor; dJ737E23.1; ECSM3; Ly68; ly-68; Lymphocyte antigen 68; matrix-remodeling-associated protein 4; matrix-remodelling associated 4; MXRA4 | |
Cd93 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.