Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD93/Early B Lin Mouse, anti-Mouse, R718, Clone: AA4.1, BD OptiBuild™

Catalog No. BDB751845 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751845 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751845 Supplier BD Biosciences Supplier No. 751845
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AA4.1 monoclonal antibody specifically recognizes the Early B Lineage antigen which is also known as CD93, AA4 antigen, Ly-68, and Complement component C1q receptor (C1qRp). This 130-140-kDa type I transmembrane glycoprotein is expressed on immature B lymphocytes in the adult bone marrow and on hematopoietic progenitors and stem cells in adult bone marrow, fetal liver, and embryonic yolk sac. Although CD93+ cells are most plentiful in adult mouse bone marrow, a smaller number of CD93+ cells which express lower CD93 levels can be detected in the adult spleen using bright fluorescent conjugates of the AA4.1 antibody or an amplified indirect immunofluorescent staining procedure. It has been observed that the staining pattern of the 493 monoclonal antibody is similar to that of the AA4.1 antibody, in that both antibodies precipitate molecules of the same molecular weight. Staining with the AA4.1 antibody is not blocked by the 493 antibody. These results suggest that the antibodies recognize separate epitopes on the same Early B Lineage antigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD93/Early B Lin
Applications Flow Cytometry
Classification Monoclonal
Clone AA4.1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.