Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD93 (Early B Lineage) Rat anti-Mouse, BB700, Clone: AA4.1, BD Optibuild™

Catalog No. BDB742187 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742187 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742187 Supplier BD Biosciences Supplier No. 742187
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The AA4.1 monoclonal antibody specifically recognizes the Early B Lineage antigen which is also known as CD93, AA4 antigen, Ly-68, and Complement component C1q receptor (C1qRp). This 130-140-kDa type I transmembrane glycoprotein is expressed on immature B lymphocytes in the adult bone marrow and on hematopoietic progenitors and stem cells in adult bone marrow, fetal liver, and embryonic yolk sac. Although CD93+ cells are most plentiful in adult mouse bone marrow, a smaller number of CD93+ cells which express lower CD93 levels can be detected in the adult spleen using bright fluorescent conjugates of the AA4.1 antibody or an amplified indirect immunofluorescent staining procedure. It has been observed that the staining pattern of the 493 monoclonal antibody is similar to that of the AA4.1 antibody, in that both antibodies precipitate molecules of the same molecular weight. Staining with the AA4.1 antibody is not blocked by the 493 antibody. These results suggest that the antibodies recognize separate epitopes on the same Early B Lineage antigen.

The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD93 (Early B Lineage)
Classification Monoclonal
Clone AA4.1
Concentration 0.2mg/mL
Conjugate Brilliant Blue 700 (BB700)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Aa4, Cd93, C1qRp, C1qr1, Ly-68, Ly68, C1q/MBL/SPA receptor
Host Species Rat
Immunogen Pre-B lymphoma 70Z/3, derived from (C57BL/6 x DBA/2)F1 mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.