Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (1032011), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10581MFV450
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow CytometrySpecifications
CD94 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Innate Immunity, Lipid and Metabolism | |
3824 | |
Store at 4C in the dark. |
Flow Cytometry | |
1032011 | |
Flow Cytometry | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
Human embryonic kidney cell HEK293-derived human CD94, Lys32-Ile179, Accession # Q13241.2 | |
RUO | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction