Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (131412), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1058H
Description
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
CD94 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Blocking Assay, CyTOF | |
131412 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
BaF3 mouse pro-B cell line transfected with human CD94 and NKG2A | |
0.5 mL | |
Innate Immunity, Lipid and Metabolism | |
3824 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction