Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (18d3), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP128121
Description
CD94 Monoclonal specifically detects CD94 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD94 | |
| Monoclonal | |
| 0.5mg/mL | |
| Flow Cytometry 1 ug/10^6 cells | |
| KLRD1 | |
| CHO cells transfected with the B6 allele of CD94 | |
| 0.25 mg | |
| Innate Immunity | |
| 3824 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 18d3 | |
| Biotin | |
| CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse CD94 receptor subunit, Mr 70 kDa | |
| Store at 4C. Do not freeze. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction