Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD94 Antibody (1H1), Novus Biologicals™
SDP

Catalog No. NBP237632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237632 Supplier Novus Biologicals Supplier No. NBP237632
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen CD94
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1H1
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor
Gene Symbols KLRD1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD94 expressed in E. Coli.
Molecular Weight of Antigen 21 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.