Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (MM0181-5F26), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212185AF700
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.Specifications
CD94 | |
Monoclonal | |
Alexa Fluor 700 | |
CAA74663.1 | |
KLRD1 | |
Human recombinant CD94 | |
0.1 mL | |
Innate Immunity | |
3824 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
MM0181-5F26 | |
Flow Cytometry | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction