Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (MM0181-5F26) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | CD94 |
---|---|
Clone | MM0181-5F26 |
Concentration | LYOPH |
Dilution | Flow Cytometry 1:10-1:1000, CyTOF-ready, Block/Neutralize |
Applications | Flow Cytometry, CyTOF, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP212185SS
![]() |
Novus Biologicals
NBP212185SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP212185
![]() |
Novus Biologicals
NBP212185 |
0.1 mg |
Each for $560.50
|
|
|||||
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD94 | |
LYOPH | |
Flow Cytometry, CyTOF, Neutralization | |
Unconjugated | |
Mouse | |
Innate Immunity | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
KLRD1 | |
IgG1 | |
Protein G purified | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
MM0181-5F26 | |
Flow Cytometry 1:10-1:1000, CyTOF-ready, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CAA74663.1 | |
3824 | |
Human recombinant CD94 | |
Primary | |
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title