Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Antibody (MM0181-5F26), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212185PEATT594
Description
CD94 Monoclonal specifically detects CD94 in Human samples. It is validated for Flow Cytometry.Specifications
CD94 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
KLRD1 | |
Human recombinant CD94 | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
MM0181-5F26 | |
Flow Cytometry | |
CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KP43, NK cell receptor | |
Mouse | |
Protein G purified | |
Innate Immunity, Lipid and Metabolism | |
3824.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction