Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD94 Monoclonal Antibody (18d3), Biotin
GREENER_CHOICE

Catalog No. PIMA528669
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528669 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528669 Supplier Invitrogen™ Supplier No. MA528669
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD94 Monoclonal Antibody for Flow

Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone 18d3
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene KLRD1
Gene Accession No. O54707
Gene Alias Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor
Gene Symbols KLRD1
Host Species Rat
Immunogen CHO cells transfected with the B6 allele of CD94.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16643
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.