Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD94 Monoclonal Antibody (18d3), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010519
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-105-19 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-105-19 Supplier Invitrogen™ Supplier No. 12094181
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 18d3 monoclonal antibody reacts with mouse CD94, a 70 kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage. DBA/2J mice are naturally CD94-deficient and do not express cell-surface CD94/NKG2A receptors, even on neonatal NK cells. Applications Reported: The 18d3 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 18d3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone 18d3
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene KLRD1
Gene Accession No. O54707
Gene Alias Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor
Gene Symbols KLRD1
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16643
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.