Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD94 Monoclonal Antibody (3G2)
GREENER_CHOICE

Catalog No. PIMA515818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515818 Supplier Invitrogen™ Supplier No. MA515818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15818 targets CD94 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15818 immunogen is purified recombinant fragment of human CD94 expressed in E. Coli. MA5-15818 detects CD94 which has a predicted molecular weight of approximately 21kDa.

Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD94
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene KLRD1
Gene Accession No. Q13241
Gene Alias Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor
Gene Symbols KLRD1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD94 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.