Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD94 Monoclonal Antibody (DX22), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 605080
Description
Positive control: Tonsil Cellular location: Membrane.
CD94 Monoclonal antibody specifically detects CD94 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
| CD94 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and 0.01% sodium azide | |
| Q13241 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| DX22 | |
| Unconjugated | |
| KLRD1 | |
| Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor | |
| A recombinant protein of human CD94 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction