Learn More
Invitrogen™ CD95 (APO-1/Fas) Monoclonal Antibody (APO-1-1), FITC, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS151FI

Description
Description: Immunohistochemistry: APO-1-1 (BMS151, IgG1) is an isotype-switch variant of the original clone APO-1-3 (IgG3). Since undesired side reactions in immunohistochemistry are minimal for IgG1 antibodies, BMS151 is especially useful for immunohistology on cryostat sections and on cytospin preparations. Flow cytometry: BMS151 is also suitable as primary antibody in staining for FACS analysis, BMS151FI can be used for direct staining of cells, BMS151BT can be used in a two-step procedure using the biotin-(strept-)avidin system. Recommended concentration for the unlabelled antibody is 1 - 2 μg/mL and a 1:20 dilution of the labelled antibodies, as determined on SKW6.4 cells. Applications Tested: Flow Cytometry. Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
FAS (Fas, CD95, APO-1) is a 46 kDa transmembrane glycoprotein that functions as a cell death receptor of the TNFR (tumor necrosis factor receptor) superfamily. FAS also exists in a soluble form that weight approximately 26kDa. Stimulation of FAS results in aggregation of its intracellular FAS-associated death domains (FADD), formation of the death-inducing signaling complex (DISC) and activation of caspases. The auto-proteolytic processing of the caspases in the complex trigger a downstream cascade that leads to apoptosis. FAS has been shown to activate NF-kappaB, MAPK3/ERK1, MAPK8/JNK, and is involved in transducing the proliferating signals in normal diploid fibroblast and T cells. At least eight alternatively spliced transcript variants encoding seven distinct isoforms have been described. The isoforms lacking the transmembrane domain may negatively regulate the apoptosis mediated by the full-length isoform. The Fas/Fas ligand system has been shown to play a role in a number of human diseases such as AIDS, hepatitis and cancer. It is also assumed that induction of apoptosis through FAS ligand pathway is predominantly involved in anti-viral immune responses.
Specifications
CD95 (APO-1/Fas) | |
Monoclonal | |
10 μL/Test | |
TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 | |
P25445 | |
FAS | |
Affinity chromatography | |
RUO | |
355 | |
4°C, store in dark | |
Liquid |
Flow Cytometry | |
APO-1-1 | |
FITC | |
FAS | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.