Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD95/Fas Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1098S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
500 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1098S1 500 μL
MS1098S0 100 μL
MS1098S 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1098S1 Supplier Epredia™ Supplier No. MS1098S1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD95/Fas Ab-3, Mouse Monoclonal Antibody.

CD95, also known as Fas, is a transmembrane glycoprotein. It is a member of the nerve growth factor receptor/tumor necrosis factor superfamily. This cell surface molecule mediates receptor-triggered apoptosis. The Fas antigen is expressed on the surface of various cell types, including activated T and B lymphocytes and T lymphoblastoid cell lines.

Host Species: Mouse

Clone: GM30

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Internal domain near the C-terminus

Immunogen: Recombinant protein corresponding to the internal domain near the C-terminal end of the Fas molecule

Molecular Weight: 48 kDa

Positive Control: Small intestine

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD95/Fas Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GM30
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the internal domain near the C-terminal end of the Fas molecule
Purification Method Small intestine
Quantity 500 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.