Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD96 Antibody (628211), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6199E
Description
CD96 Monoclonal antibody specifically detects CD96 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD96 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
628211 | |
Flow Cytometry, CyTOF-ready | |
CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILEincreased late expression, T-cell surface protein tactile | |
Mouse myeloma cell line NS0-derived recombinant human CD96, Val22-Asp501, Accession # NP_005807 | |
0.1 mL | |
Immunology | |
10225 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction