Learn More
Invitrogen™ CD96 Chimeric Recombinant Rabbit Monoclonal Antibody (6A6)

Description
Specificity: This antibody is specific for murine CD96, and recognises an epitope in the first Ig domain.
Specifications
Specifications
| Antigen | CD96 Chimeric |
| Applications | ELISA, Flow Cytometry, Inhibition Assays |
| Classification | Recombinant Monoclonal |
| Clone | 6A6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Cd96 |
| Gene Accession No. | Q3U0X8 |
| Gene Alias | 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.