Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD96 Monoclonal Antibody (628211)
GREENER_CHOICE

Catalog No. PIMA524280
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524280 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524280 Supplier Invitrogen™ Supplier No. MA524280
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD96
Applications Flow Cytometry
Classification Monoclonal
Clone 628211
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Cd96
Gene Accession No. P40200
Gene Alias 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile
Gene Symbols Cd96
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD96 Val22-Asp501.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10225
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.