Learn More
Invitrogen™ CD96 Monoclonal Antibody (630612)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | CD96 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 630612 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Cd96 |
| Gene Accession No. | Q3U0X8 |
| Gene Alias | 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.