Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD96 Mouse anti-Human, Brilliant Violet 421, Clone: 6F9, BD Optibuild™

Catalog No. BDB742794 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742794 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742794 Supplier BD Biosciences Supplier No. 742794
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6F9 monoclonal antibody specifically binds to human CD96, also known as TACTILE (T cell activation increased late expression). CD96 is a type I transmembrane glycoprotein and member of the Ig superfamily. CD96 is expressed at low levels on resting natural killer (NK) cells and T cells and at high levels on activated NK and T cells. CD96 is also expressed on some T-cell leukemia and acute myeloid leukemia cells. CD96 may serve as a marker for acute myelogenous leukemia stem cells. CD96 plays a role in the adhesive interactions of activated NK and T cells during immune responses. CD96 binds to the poliovirus receptor (CD155) that is highly expressed by some tumor cells. CD155-mediated ligation of CD96 can induce NK cell-mediated cytotoxicity. CD96-mediated uptake of CD155 may adversely affect NK cells and thus reduce their effectiveness in anti-tumor responses

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD96
Classification Monoclonal
Clone 6F9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TACTILE, T cell activation increased late expression
Host Species Mouse
Immunogen Human CD96 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.