Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody (3A7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CD97 |
---|---|
Clone | 3A7 |
Dilution | ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
CD97 Monoclonal antibody specifically detects CD97 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
CD97 | |
ELISA, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence 1:30-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Synthesized peptide [UniProt P48960] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3A7 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
GPCR, Immunology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
976 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title