Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299764IR
Description
CD97 Polyclonal specifically detects CD97 in Human samples. It is validated for Western Blot, ELISA.Specifications
CD97 | |
Polyclonal | |
Western Blot, ELISA | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Produced in rabbits immunized with purified, recombinant CD97 (NP_001775.2; Met 1-Gln 398). | |
0.1 mL | |
GPCR, Immunology | |
976 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction