Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody (MEM-180), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500425AF647
Description
CD97 Monoclonal specifically detects CD97 in Human samples. It is validated for Flow Cytometry.Specifications
CD97 | |
Monoclonal | |
Alexa Fluor 647 | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody MEM-180 recognizes an unique epitope on CD97, a 75-85 kDa surface glycoprotein of G-protein-coupled receptor family, expressed on activated B and T lymphocytes, monocytes/macrophages, dendritic cells and granulocytes. HLDA VI; WS Code BP 415 HLDA VI; WS Code NL N-L023 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
MEM-180 | |
Flow Cytometry | |
ADGRE5 | |
PHA-activated peripheral blood cells | |
0.1 mL | |
GPCR, Immunology | |
976 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction