Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody (MEM-180), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500425MFV450
Description
CD97 Monoclonal antibody specifically detects CD97 in Human samples. It is validated for Flow CytometrySpecifications
CD97 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MEM-180 | |
Flow Cytometry | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
PHA-activated peripheral blood cells | |
0.1 mL | |
GPCR, Immunology | |
976 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction