Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Isoform 1 Antibody (587702), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3734B
Description
CD97 Isoform 1 Monoclonal antibody specifically detects CD97 Isoform 1 in Mouse samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
CD97 Isoform 1 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
976 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Blocking Assay, CyTOF | |
587702 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
CD97, TM7LN1 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse CD97v2, Gln24-His384, Accession # AAH06676 | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction