Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Mouse anti-Human, Brilliant Violet 510, Clone: VIM3b, BD Optibuild™

Catalog No. BDB742446 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742446 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742446 Supplier BD Biosciences Supplier No. 742446
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The VIM3b monoclonal antibody recognizes CD97. CD97 is a seven-span transmembrane protein that belongs to the G-protein coupled receptor superfamily. CD97 is encoded by ADGRE5 (adhesion G protein-coupled receptor E5). CD97 is highly expressed on monocytes and granulocytes. It is also expressed on dendritic cells, T cells, B cells, NK cells, and smooth muscle cells. CD97 expression is upregulated on activated T cells. CD97 is also expressed on several cell lines including HL-60, THP-1, and K562. CD97 reportedly plays roles in leucocyte signaling, adhesion, and migration. The VIM3b antibody works in Western blotting and immunoprecipitation. The antibody recognizes proteins of 74, 80 and 89 kDa. CD55 and chondroitin sulfate are ligands for CD97.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD97
Classification Monoclonal
Clone VIM3b
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ADGRE5: BL-KDD/F12, TL7LN1
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.