Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD97 Mouse anti-Human, BUV395, Clone: VIM3b, BD Optibuild™

Catalog No. BDB742451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742451 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742451 Supplier BD Biosciences Supplier No. 742451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The VIM3b monoclonal antibody recognizes CD97. CD97 is a seven-span transmembrane protein that belongs to the G-protein coupled receptor superfamily. CD97 is encoded by ADGRE5 (adhesion G protein-coupled receptor E5). CD97 is highly expressed on monocytes and granulocytes. It is also expressed on dendritic cells, T cells, B cells, NK cells, and smooth muscle cells. CD97 expression is upregulated on activated T cells. CD97 is also expressed on several cell lines including HL-60, THP-1, and K562. CD97 reportedly plays roles in leucocyte signaling, adhesion, and migration. The VIM3b antibody works in Western blotting and immunoprecipitation. The antibody recognizes proteins of 74, 80 and 89 kDa. CD55 and chondroitin sulfate are ligands for CD97.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD97
Classification Monoclonal
Clone VIM3b
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ADGRE5: BL-KDD/F12, TL7LN1
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.