Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD97 Recombinant Rabbit Monoclonal Antibody (3E5L8)
GREENER_CHOICE

Catalog No. FEMA551373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551373 Supplier Invitrogen™ Supplier No. MA551373
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: QDSRGCARWC PQNSSCVNAT ACRCNPGFSS FSEIITTPTE TCDDINECAT PSKVSCGKFS DCWNTEGSYD CVCSPGYEPV SGAKTFKNES ENTCQDVDEC QQNPRLCKSY GTCVNTLGSY TCQCLPGFKF IPEDPKVCTD VNECTSGQNP CHSSTHCLNN VGSYQCRCRP GWQPIPGSPN GPNNTVCEDV DECSSGQHQC DSSTVCFNTV GSYSCRCRPG WKPRHGIPNN QKDTVCEDMT FSTWTPPPGV HSQTLSRFFD KVQDLGRDSK TSSAEVTIQN VIKLVDELME APGDVEALAP PVRHLIATQL LSNLEDIMRI LAKSLPKGPF TYISPSNTEL TLMIQERGDK NVTMGQSSAR MKLNWAVAAG AEDPGPAV.

CD97 is a G-protein-coupled seven-span transmembrane adhesive receptor that is constitutively expressed on granulocytes and monocytes and rapidly upregulated on T and B cells upon activation. CD97 is produced in alternatively spliced forms and its cellular ligand is CD55 (DAF), which protects various cell types from complement-mediated damage. Interaction of CD97 on leukocytes and CD55 on vessel cells probably facilitate leukocyte activation and migration into the tissues, similarly, CD97 seems to play a role in tumour migration and invasiveness. CD97 is involved in T cell regulation and peripheral granulocyte homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD97
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3E5L8
Concentration 0.072 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ADGRE5
Gene Accession No. P48960
Gene Alias AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1
Gene Symbols ADGRE5, CD97
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-398 of human CD97 (NP_510966.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.